Learn More
Biomatik Corporation Human Fibrinogen Gamma Chain (Fgg) Elisa Kit
Supplier:  Biomatik Corporation EKC33683
Encompass_Preferred
Human Fibrinogen Gamma Chain (Fgg) Elisa Kit for detecting Fibrinogen Gamma Chain in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8000ng/ml. Sensitivity: 31.25ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P02679; FGG. Target Synonyms: FGG; FIBG_HUMAN; Fibrinogen gamma chain; Fibrinogen gamma polypeptide; fibrinogen gamma-b chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7415