Learn More
Biomatik Corporation Human Ghrelin (Ghrl) Elisa Kit
Supplier:  Biomatik Corporation EKC33811
Encompass_Preferred
Human Ghrelin (Ghrl) Elisa Kit for detecting Ghrelin/Obestatin Prepropeptide in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40pg/ml. Sensitivity: 0.156pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9UBU3. Target Synonyms: Appetite regulating hormone; Ghrelin 27; Ghrelin 28; Ghrelin and obestatin prepropeptide; Ghrelin; Ghrelin/obestatin prepropeptide; ghrl; GHRL_HUMAN; Growth hormone releasing peptide; Growth hormone secretagogue; Growth hormone-releasing peptide; M46 protein; Motilin related peptide; Motilin-related peptide; MTLRP; Obestatin; Protein M46. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7422