Learn More
Biomatik Corporation Bovine fibroblast growth factor 21 (FGF21) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC31627
Encompass_Preferred
The Bovine fibroblast growth factor 21 (FGF21) ELISA Kit allows for quantitative determination of fibroblast growth factor 21 in serum, plasma, tissue homogenates. Species Reactivity: Bovine. Detection Range: 62.5 pg/mL-4000 pg/mL. Sensitivity: 15.6 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-7492