Learn More
Biomatik Corporation Human Hla Class Ii Histocompatibility Antigen Gamma Chain (Cd74) Elisa Kit
Supplier:  Biomatik Corporation EKC34055
Encompass_Preferred
Human Hla Class Ii Histocompatibility Antigen Gamma Chain (Cd74) Elisa Kit for detecting Cd74 Molecule, Major Histocompatibility Complex, Class Ii Invariant Chain in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.9-60ng/ml. Sensitivity: 0.225ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04233; CD74. Target Synonyms: CD74; DHLAG; HLA class II histocompatibility antigen gamma chain; HLA-DR antigens-associated invariant chain; Ia antigen-associated invariant chain; Ii; CD antigen CD74. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7432