Learn More
Biomatik Corporation Human Inducible Nitric Oxide Synthase, Inos Elisa Kit
Supplier:  Biomatik Corporation EKC34125
Encompass_Preferred
Human Inducible Nitric Oxide Synthase, Inos Elisa Kit for detecting Nitric Oxide Synthase 2, Inducible in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.9-60IU/ml. Sensitivity: 0.225IU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P35228; NOS2. Target Synonyms: HEP-NOS; Hepatocyte NOS; HEPNOS; inducible; Inducible nitric oxide synthase; Inducible NO synthase; Inducible NOS; iNOS; MAC NOS; Macrophage NOS; Nitric oxide synthase 2 inducible; Nitric oxide synthase 2 inducible macrophage; nitric oxide synthase 2A (inducible; hepatocytes); Nitric oxide synthase; Nitric oxide synthase inducible; nitric oxide synthase; macrophage; NOS 2; NOS; Nos II; NOS type II; nos2; NOS2_HUMAN; NOS2A; NOS2A; Inducible; Hepatocyte; Peptidyl-cysteine S-nitrosylase NOS2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7441