Learn More
Biomatik Corporation Human Insulin-Like Growth Factor-Binding Protein 7 (Igfbp7/Mac25/Psf) Elisa Kit
Supplier:  Biomatik Corporation EKC34149
Encompass_Preferred
Human Insulin-Like Growth Factor-Binding Protein 7 (Igfbp7/Mac25/Psf) Elisa Kit for detecting Insulin-Like Growth Factor Binding Protein 7 in serum, plasma, homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q16270; IGFBP7. Target Synonyms: AGM; Angiomodulin; FSTL2; IBP-7; IBP7; IBP7_HUMAN; IGF binding protein 7; IGF-binding protein 7; IGFBP rP1; IGFBP-7; IGFBP-rP1; IGFBP7; IGFBPRP1; Insulin like growth factor binding protein 7; Insulin-like growth factor-binding protein 7; MAC25; MAC25 protein; PGI2 stimulating factor; PGI2-stimulating factor; Prostacyclin stimulating factor; Prostacyclin-stimulating factor; PSF; RAMSVPS; TAF; Tumor-derived adhesion factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7444