Learn More
Biomatik Corporation Human Lactoferrin, Ltf/Lf Elisa Kit
Supplier:  Biomatik Corporation EKC34363
Encompass_Preferred
Human Lactoferrin, Ltf/Lf Elisa Kit for detecting Lactotransferrin in breast milk. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ug/ml. Sensitivity: 0.78ug/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P02788; LTF. Target Synonyms: GIG12; Growth inhibiting protein 12; HLF2; Kaliocin 1; Lactoferrin; Lactoferroxin-C; Lactotransferrin; LF; LTF; Neutrophil lactoferrin; Talalactoferrin; TRFL_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7465