Learn More
Biomatik Corporation Human Mammalian Target Of Rapamyein, Mtor Elisa Kit
Supplier:  Biomatik Corporation EKC34534
Encompass_Preferred
Human Mammalian Target Of Rapamyein, Mtor Elisa Kit for detecting Mammalian Target Of Rapamyein, Mtor in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100pg/ml. Sensitivity: 0.39pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P42345; MTOR. Target Synonyms: dJ576K7.1 (FK506 binding protein 12 rapamycin associated protein 1); FK506 binding protein 12 rapamycin associated protein 1; FK506 binding protein 12 rapamycin associated protein 2; FK506 binding protein 12 rapamycin complex associated protein 1; FK506-binding protein 12-rapamycin complex-associated protein 1; FKBP rapamycin associated protein; FKBP12 rapamycin complex associated protein; FKBP12-rapamycin complex-associated protein 1; FKBP12-rapamycin complex-associated protein; FLJ44809; FRAP; FRAP1; Mammalian target of rapamycin; Mechanistic target of rapamycin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7477