Learn More
Biomatik Corporation Human Midkine, Mk Elisa Kit
Supplier:  Biomatik Corporation EKC34615
Encompass_Preferred
Human Midkine, Mk Elisa Kit for detecting Midkine (Neurite Growth-Promoting Factor 2) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P21741; MDK. Target Synonyms: Amphiregulin associated protein; Amphiregulin-associated protein; ARAP; FLJ27379; Mdk; Midgestation and kidney protein; Midkine; MK 1; MK; MK_HUMAN; MK1; NEGF 2; NEGF2; Neurite growth promoting factor 2; Neurite outgrowth promoting protein; Neurite outgrowth-promoting factor 2; Neurite outgrowth-promoting protein; Retinoic acid inducible factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7482