Learn More
Biomatik Corporation Human Myeloid Cell Surface Antigen Cd33 (Cd33) Elisa Kit
Supplier:  Biomatik Corporation EKC34698
Encompass_Preferred
Human Myeloid Cell Surface Antigen Cd33 (Cd33) Elisa Kit for detecting Cd33 Molecule in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8000pg/ml. Sensitivity: 31.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P20138; CD33. Target Synonyms: CD33; SIGLEC3; Myeloid cell surface antigen CD33; Sialic acid-binding Ig-like lectin 3; Siglec-3; gp67; CD antigen CD33. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7486