Learn More
Biomatik Corporation Human Neuropeptide Y (Npy) Elisa Kit
Catalog No. 502287494
Encompass_Preferred
Human Neuropeptide Y (Npy) Elisa Kit for detecting Neuropeptide Y in serum, plasma, cell culture supernates, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50pg/ml. Sensitivity: 0.195pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01303; NPY. Target Synonyms: C-flanking peptide of NPY; CPON; Neuropeptide tyrosine; Neuropeptide Y precursor; NPY; NPY_HUMAN; Pro neuropeptide Y; PYY 4; PYY4; Y Neuropeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7494 Supplier Biomatik Corporation Supplier No. EKC34803
Add to Cart
Add to Cart
Human Neuropeptide Y (Npy) Elisa Kit for detecting Neuropeptide Y in serum, plasma, cell culture supernates, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50pg/ml. Sensitivity: 0.195pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01303; NPY. Target Synonyms: C-flanking peptide of NPY; CPON; Neuropeptide tyrosine; Neuropeptide Y precursor; NPY; NPY_HUMAN; Pro neuropeptide Y; PYY 4; PYY4; Y Neuropeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.