Learn More
Biomatik Corporation Human Omentin Elisa Kit
Supplier:  Biomatik Corporation EKC34881
Encompass_Preferred
Human Omentin Elisa Kit for detecting Intelectin 1 (Galactofuranose Binding) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100pg/ml. Sensitivity: 0.39pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8WWA0; ITLN1. Target Synonyms: Endothelial lectin HL 1; Endothelial lectin HL-1; FLJ20022; Galactofuranose binding lectin; Galactofuranose-binding lectin; hIntL; HL 1; HL-1; HL1; Intelectin; Intelectin 1; Intelectin-1; Intestinal lactoferrin receptor; INTL; ITLN; ITLN-1; ITLN1; ITLN1_HUMAN; LFR; Omentin; OTTHUMP00000027882. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7498