Learn More
Biomatik Corporation Human Oxytocin, Ot Elisa Kit
Supplier:  Biomatik Corporation EKC34910
Encompass_Preferred
Human Oxytocin, Ot Elisa Kit for detecting Oxytocin, Ot in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 8-400uIU/ml. Sensitivity: 10uIU/mL. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7504