Learn More
Biomatik Corporation Human Pepsin,PG ELISA Kit
Supplier:  Biomatik Corporation EKC34940
Encompass_Preferred
Sample Type: serum, plasma, urine, tissue homogenates. Species Reactivity: Homo sapiens (Human). Detection Range: 31.25 pg/mL-2000 pg/mL. Sensitivity: 7.81 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P0DJD8. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7509