Learn More
Biomatik Corporation Human Phospholipidase A2, Pla2 Elisa Kit
Supplier:  Biomatik Corporation EKC35015
Encompass_Preferred
Human Phospholipidase A2, Pla2 Elisa Kit for detecting Phospholipase A2, Group Ib (Pancreas) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.45-30ng/ml. Sensitivity: 0.11ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04054; PLA2G1B. Target Synonyms: Group IB phospholipase A2; Group IB secretory phospholipase A2; MGC119834; MGC119835; PA21B_HUMAN; Phosphatidylcholine 2 acylhydrolase; Phosphatidylcholine 2-acylhydrolase 1B; Phospholipase A2; Phospholipase A2 group IB (pancreas); Phospholipase A2 group IB; PLA 2; PLA2; PLA2A; PLA2G1B; PPLA 2; PPLA2; sPLA2IB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7513