Learn More
Biomatik Corporation Human plasma kallikrein(KLKB1) ELISA kit
Supplier:  Biomatik Corporation EKC35033
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 9.38 mU/mL-600 mU/mL. Sensitivity: 2.34 mU/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P03952. Target Synonyms: KLK3, PPK, Fletcher factor|kininogenin|plasma kallikrein B1|plasma kallikrein heavy chain|plasma kallikrein light chain. Restriction: For Research Use Only.
Catalog No. 50-228-7518