Learn More
Biomatik Corporation Human Prothrombin Fragment 1+2, F1+2 Elisa Kit
Supplier:  Biomatik Corporation EKC35279
Encompass_Preferred
Human Prothrombin Fragment 1+2, F1+2 Elisa Kit for detecting Prothrombin Fragment 1+2, F1+2 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.03-2nmol/L. Sensitivity: 0.007nmol/l. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7532