Learn More
Biomatik Corporation Human Pseudomonas Exotoxin A,PEA ELISA Kit
Supplier:  Biomatik Corporation EKC35294
Encompass_Preferred
Sample Type: serum, saliva, tissue homogenates. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7534