Learn More
Biomatik Corporation Human Rho Gdp-Dissociation Inhibitor 2 (Arhgdib) Elisa Kit
Supplier:  Biomatik Corporation EKC35396
Encompass_Preferred
Human Rho Gdp-Dissociation Inhibitor 2 (Arhgdib) Elisa Kit for detecting Rho Gdp Dissociation Inhibitor (Gdi) Beta in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P52566; ARHGDIB. Target Synonyms: Arhgdib; D4; D4 GDP dissociation inhibitor; GDIA 2; GDIA2; GDID 4; GDID4; GDIR2_HUMAN; GDP dissociation inhibitor D4; LY GDI; Ly-GDI; LYGDI; MGC108926; RAP1GN1; Rho GDI 2; Rho GDI beta; Rho GDI2; Rho GDP dissociation inhibitor (GDI) beta; Rho GDP dissociation inhibitor 2; Rho GDP dissociation inhibitor beta; Rho GDP-dissociation inhibitor 2; Rho-GDI beta; RhoGDI2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7539