Learn More
Biomatik Corporation Human Sex Hormone-Binding Globulin, Shbg Elisa Kit
Supplier:  Biomatik Corporation EKC35545
Encompass_Preferred
Human Sex Hormone-Binding Globulin, Shbg Elisa Kit for detecting Sex Hormone-Binding Globulin in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100pmol/l. Sensitivity: 0.39pmol/l. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04278; SHBG. Target Synonyms: ABP; Androgen binding protein; SBP; Sex hormone-binding globulin; Sex steroid binding protein; Sex steroid-binding protein; SHBG; SHBG_HUMAN; TeBG; Testis specific androgen binding protein; Testis-specific androgen-binding protein; Testosterone binding beta globulin; Testosterone estradiol binding globulin; Testosterone estrogen binding globulin; Testosterone-estradiol-binding globulin; Testosterone-estrogen-binding globulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7543