Learn More
Biomatik Corporation Human Telomerase, Te Elisa Kit
Supplier:  Biomatik Corporation EKC35734
Encompass_Preferred
Human Telomerase, Te Elisa Kit for detecting Telomerase, Te in serum, cell culture supernates, urine, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7549