Learn More
Biomatik Corporation Human α2-Antiplasmin,α2-AP ELISA Kit
Supplier:  Biomatik Corporation EKC36096
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P08697. Target Synonyms: A2AP, AAP, ALPHA-2-PI, API, PLI, OTTHUMP00000115656|alpha-2-antiplasmin|alpha-2-plasmin inhibitor|serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived. Restriction: For Research Use Only.
Catalog No. 50-228-7554