Learn More
Biomatik Corporation Monkey C-C motif chemokine 17(CCL17) ELISA kit
Supplier:  Biomatik Corporation EKC36143
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Monkey. Detection Range: 39 pg/mL-2500 pg/mL. Sensitivity: 9.8 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q8HYP9. Target Synonyms: A-152E5.3, ABCD-2, MGC138271, MGC138273, SCYA17, TARC, T cell-directed CC chemokine|small inducible cytokine A17|small inducible cytokine subfamily A (Cys-Cys), member 17|thymus and activation-regul. Restriction: For Research Use Only.
Catalog No. 50-228-7559