Learn More
Biomatik Corporation Mouse Acetyl-Coa Elisa Kit
Supplier:  Biomatik Corporation EKC36240
Encompass_Preferred
Mouse Acetyl-Coa Elisa Kit for detecting Acetyl-Coa in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.195ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7567