Learn More
Biomatik Corporation Mouse Apolipoprotein A-I-binding protein(APOA1BP) ELISA kit
Supplier:  Biomatik Corporation EKC36350
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 9.375 pg/mL-600 pg/mL. Sensitivity: 2.34 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q8K4Z3. Target Synonyms: RP11-284F21.3, AIBP, MGC119143, MGC119144, MGC119145, YJEFN1, apoA-I binding protein. Restriction: For Research Use Only.
Catalog No. 50-228-7574