Learn More
Biomatik Corporation Mouse C-C chemokine receptor type 2(CCR2) ELISA kit
Supplier:  Biomatik Corporation EKC36500
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P51683. Target Synonyms: CC-CKR-2, CCR2A, CCR2B, CD192, CKR2, CKR2A, CKR2B, CMKBR2, MCP-1-R, MCP-1 receptor|chemokine (C-C) receptor 2|monocyte chemoattractant protein 1 receptor|monocyte chemotactic protein 1 receptor. Restriction: For Research Use Only.
Catalog No. 50-228-7579