Learn More
Biomatik Corporation Mouse Glucagon (GC) ELISA Kit
Supplier:  Biomatik Corporation EKC36943
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 1.56 pg/mL-100 pg/mL. Sensitivity: 0.39 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P55095. Target Synonyms: GLP1, GLP2, GRPP, glicentin-related polypeptide|glucagon-like peptide 1|glucagon-like peptide 2. Restriction: For Research Use Only.
Catalog No. 50-228-7595