Learn More
Biomatik Corporation Mouse Interferon α1,IFN-α1 ELISA Kit
Supplier:  Biomatik Corporation EKC37142
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 3.12 pg/mL-200 pg/mL. Sensitivity: 0.78 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P01572. Target Synonyms: IFN-alpha 1,interferon alpha 1. Restriction: For Research Use Only.
Catalog No. 50-228-7602