Learn More
Biomatik Corporation Mouse monocyte chemotactic protein 3,MCP-3 ELISA kit
Supplier:  Biomatik Corporation EKC37405
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 1.56 pg/mL-100 pg/mL. Sensitivity: 0.39 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q03366. Target Synonyms: FIC, MARC, MCP-3, MCP3, MGC138463, MGC138465, NC28, SCYA6, SCYA7, monocyte chemoattractant protein 3|monocyte chemotactic protein 3|small inducible cytokine A7 (monocyte chemotactic protein 3). Restriction: For Research Use Only.
Catalog No. 50-228-7612