Learn More
Biomatik Corporation Mouse Oncostatin-M, Osm Elisa Kit
Supplier:  Biomatik Corporation EKC37519
Encompass_Preferred
Mouse Oncostatin-M, Osm Elisa Kit for detecting Oncostatin M in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P53347; Osm. Target Synonyms: Osm; Oncostatin-M; OSM. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7615