Learn More
Biomatik Corporation Mouse platelet-derived growth factor-BB,PDGF-BB ELISA Kit
Supplier:  Biomatik Corporation EKC37601
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.18 ng/mL-12 ng/mL. Sensitivity: 0.047 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7619