Learn More
Biomatik Corporation Pig Epithelial growth factor,EGF ELISA Kit
Supplier:  Biomatik Corporation EKC38169
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Sus scrofa (Pig). Detection Range: 250 pg/mL-10000 pg/mL. Sensitivity: 125 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: F1S137. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7631