Learn More
Biomatik Corporation Pig Haptoglobin,Hpt/HP ELISA Kit
Supplier:  Biomatik Corporation EKC38203
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Sus scrofa (Pig). Detection Range: 1.56 μg/mL-100 μg/mL. Sensitivity: 0.39 μg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q8SPS7. Target Synonyms: BP, HP2-ALPHA-2, HPA1S, MGC111141, binding peptide|haptoglobin alpha(1S)-beta|haptoglobin alpha(2FS)-beta|haptoglobin, alpha polypeptide|haptoglobin, beta polypeptide. Restriction: For Research Use Only.
Catalog No. 50-228-7633