Learn More
Biomatik Corporation Rat Angiotensin converting enzyme 2, ACE2 ELISA Kit
Supplier:  Biomatik Corporation EKC38667
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.02 ng/ml - 20 ng/ml. Sensitivity: 0.02 ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q5EGZ1. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7647