Learn More
Biomatik Corporation Rat Cross Linked N-Telopeptide Of Type I Collagen, Ntx Elisa Kit
Supplier:  Biomatik Corporation EKC38914
Encompass_Preferred
Rat Cross Linked N-Telopeptide Of Type I Collagen, Ntx Elisa Kit for detecting Cross Linked N-Telopeptide Of Type I Collagen, Ntx in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 4.7-300nM BCE. Sensitivity: 1.17nM BCE. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: cross linked N-telopeptide of type 1 collagen; NTX; cross linked N-telopeptide of type I collagen; NTX. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7654