Learn More
Biomatik Corporation Rat Gastrin Elisa Kit
Supplier:  Biomatik Corporation EKC39089
Encompass_Preferred
Rat Gastrin Elisa Kit for detecting Gastrin in serum, plasma, cell culture supernates , tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20pg/ml. Sensitivity: 0.078pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04563; Gast. Target Synonyms: Gast; Gas; Gastrin [Cleaved into: Big gastrin; Gastrin-34; G34); Gastrin]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7660