Learn More
Biomatik Corporation Rat Neuronal Nitric Oxide Synthase (Nnos/Nos1) Elisa Kit
Supplier:  Biomatik Corporation EKC39476
Encompass_Preferred
Rat Neuronal Nitric Oxide Synthase (Nnos/Nos1) Elisa Kit for detecting Nitric Oxide Synthase 1 (Neuronal) in serum, plasma, cell culture supernates, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000mIU/ml. Sensitivity: 7.81mlU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P29476; Nos1. Target Synonyms: Nos1; BnosNitric oxide synthase; brain; EC 1.14.13.39; BNOS; Constitutive NOS; NC-NOS; NOS type I; Neuronal NOS; N-NOS; nNOS; Peptidyl-cysteine S-nitrosylase NOS1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7671