Learn More
Biomatik Corporation Rat Prealbumin, Pa Elisa Kit
Supplier:  Biomatik Corporation EKC39572
Encompass_Preferred
Rat Prealbumin, Pa Elisa Kit for detecting Prealbumin, Pa in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 375-6000ng/ml. Sensitivity: 375ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P02767; Ttr. Target Synonyms: Ttr; Tt; Transthyretin; Prealbumin; TBPA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7677