Learn More
Biomatik Corporation Sheep neutrophil gelatinase-associated lipocalin,NGAL ELISA Kit
Supplier:  Biomatik Corporation EKC39977
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Ovis aries (Sheep). Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7687