Learn More
Biomatik Corporation Human Fractalkine, Fk Elisa Kit
Supplier:  Biomatik Corporation EKC40045
Encompass_Preferred
Human Fractalkine, Fk Elisa Kit for detecting Chemokine (C-X3-C Motif) Ligand 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P78423; CX3CL1. Target Synonyms: A 152E5.2; AB030188; ABCD 3; ABCD3; AI848747; C-X3-C motif chemokine 1; C3Xkine; Chemokine (C-X3-C motif) ligand 1; Chemokine C X3 C motif ligand 1; Chemokine CX3C Motif Ligand 1; CX3C membrane anchored chemokine; CX3C membrane-anchored chemokine; Cx3cl1; CXC 3; CXC3; CXC3C; D8Bwg0439e; FKN; Fractalkine; Neurotactin; NTN; NTT; Processed fractalkine; SCYD 1; SCYD1; Small inducible cytokine D1; Small inducible cytokine subfamily D (Cys X3 Cys) member 1; small inducible cytokine subfamily D (Cys-X3-Cys); member 1 (fractalkine; neurotactin). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7695