Learn More
Biomatik Corporation Mouse MMP-1 (Matrix Metalloproteinase 1) ELISA Kit
Supplier:  Biomatik Corporation EKF58008
Encompass_Preferred
Mouse MMP-1 (Matrix Metalloproteinase 1) ELISA Kit for detecting MMP-1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 125-8000pg/ml. Sensitivity: 75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9EPL5. Target Synonyms: MMP-1; Matrix metallopeptidase 1; CLGmatrix metalloprotease 1; CLGN; Fibroblast collagenase; interstitial collagenase; matrix metalloproteinase 1; interstitial collagenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8005