Learn More
Biomatik Corporation Rat Ache (Acetylcholinesterase) ELISA Kit
Supplier:  Biomatik Corporation EKF58039
Encompass_Preferred
Rat Ache (Acetylcholinesterase) ELISA Kit for detecting Ache in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P37136. Target Synonyms: ACHE; Acetylcholinesterase; ACEE; ARACHE; N-ACHE; YT; Acetylcholinesterase YT blood group; Apoptosis related acetylcholinesterase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8019