Learn More
Biomatik Corporation Human HSP72 (Heat Shock Protein 72) ELISA Kit
Supplier:  Biomatik Corporation EKF58059
Encompass_Preferred
Human HSP72 (Heat Shock Protein 72) ELISA Kit for detecting HSP72 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P0DMV8. Target Synonyms: HSP-72. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8029