Learn More
Biomatik Corporation Human IgA1 (Immunoglobulin A1) ELISA Kit
Supplier:  Biomatik Corporation EKF58132
Encompass_Preferred
Human IgA1 (Immunoglobulin A1) ELISA Kit for detecting IgA1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.5-160ng/ml. Sensitivity: 1.5ng/ml. Format: 96-Well Plate. Assay Type: HRP Antibody, Sandwich ELISA. Assay Time: 3h, 30 min. Accession Number: P01876. Target Synonyms: IgA1; Immunoglobulin A1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8040