Learn More
Biomatik Corporation Mouse ANA (Anti-nuclear Antibody) ELISA Kit
Supplier:  Biomatik Corporation EKF58166
Encompass_Preferred
Mouse ANA (Anti-nuclear Antibody) ELISA Kit for detecting ANA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.469-30ng/ml. Sensitivity: 0.281ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antigen. Assay Time: 3h, 30 min. Target Synonyms: Anti-nuclear Antibody. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8052