Learn More
Biomatik Corporation Human TPMT (Thiopurine S-methyltransferase) ELISA Kit
Supplier:  Biomatik Corporation EKF58177
Encompass_Preferred
Human TPMT (Thiopurine S-methyltransferase) ELISA Kit for detecting TPMT in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.125-200ng/ml. Sensitivity: 1.875ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P51580. Target Synonyms: TPMT; Thiopurine S-methyltransferase; Thiopurine methyltransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8056