Learn More
Biomatik Corporation Mouse KYN (Kynurenine) ELISA Kit
Supplier:  Biomatik Corporation EKF58197
Encompass_Preferred
Mouse KYN (Kynurenine) ELISA Kit for detecting KYN in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.813-500pmol/ml. Sensitivity: 4.688pmol/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: Kynurenine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8061