Learn More
Biomatik Corporation Human anti-NMDAR Antibody (anti-N-methyl-D-aspartic Acid receptor Antibody) ELISA Kit
Supplier:  Biomatik Corporation EKF58220
Encompass_Preferred
Human anti-NMDAR Antibody (anti-N-methyl-D-aspartic Acid receptor Antibody) ELISA Kit for detecting anti-NMDAR Antibody in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antigen. Assay Time: 3h, 30 min. Accession Number: Q12879; Q13224. Target Synonyms: anti-NMDAR Antibody. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8067