Learn More
Biomatik Corporation Mouse HS (Heparan Sulfate) ELISA Kit
Supplier:  Biomatik Corporation EKF58232
Encompass_Preferred
Mouse HS (Heparan Sulfate) ELISA Kit for detecting Heparan Sulfate in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000ng/ml. Sensitivity: 18.75ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: Heparan Sulfate. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8072