Learn More
Biomatik Corporation Mouse Irisin ELISA Kit
Supplier:  Biomatik Corporation EKF58236
Encompass_Preferred
Mouse Irisin ELISA Kit for detecting Irisin in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8K4Z2. Target Synonyms: Irisin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8074